1-Heptanesulfonic acid sodium salt, (max. 2% H₂O) 99% (dry weight)

Supplier: Thermo Scientific Chemicals
Warning

A10917-14 A10917-22 A10917-06
CAA10917-14EA 235.42 CAD
CAA10917-14 CAA10917-22 CAAAA10917-06
1-Heptanesulfonic acid sodium salt, (max. 2% H₂O) 99% (dry weight)
1-Heptanesulphonic acid sodium salt

HPLC reagent used for ion-association in analyses of peptides and proteins


Purity: 99% CAS No.: 22767-50-6 Molecular Formula: CH3(CH2)6SO3Na Formula Weight: 202.24 Odor: Odorless Melting Point: 300° Beilstein Reference: 3731762 MDL No.: MFCD00007543

Formula: H₃C(CH₂)₅CH₂SO₃Na
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00007543
CAS Number: 22767-50-6
EINECS: 245-210-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=95c09a5691140bab7446d88919f01593e8f0b9b2&vid=7c237cde5db01d0567ff19964ca3287434747480&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR