Sodium tetraphenylborate 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A10909-06
A10909-14
A10909-22
CAAAA10909-06EA
66.36
CAD
CAAAA10909-06
CAAAA10909-14
CAAAA10909-22
Sodium tetraphenylborate 99%
Sodium tetraphenylborate
MDL: MFCD00011494
Beilstein Registry No.: 3599783
Purity: 99%
CAS No.: 143-66-8
Molecular Formula: C24H20BNa
Formula Weight: 342.23
pH: 8
Melting Point:
Formula:
(C₆H₅)₄BNa Melting Pt: >300 °C Density: 1.15 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011494 CAS Number: 143-66-8 EINECS: 205-605-5 UN: 2811 ADR: 6.1,III Merck Index: 14,8690 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...