Ethyl-(S)-(-)-lactate 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Ethyl (S)-(-)-2-hydroxypropionate, Ethyl L(-)-lactate, (S)-Ethyl 2-hydroxypropanoate, (S)-(-)-Ethyl lactate

A10900-0E A10900-36
CAAAA10900-0EEA 606.72 CAD
CAAAA10900-0E CAAAA10900-36
Ethyl-(S)-(-)-lactate 99%
Ethyl (S)-(-)-lactate

MDL: MFCD00065359 Beilstein Registry No.: 1720839 Fieser: 12,226 17,135 Optical Rotation: -10.7°C (neat)


Purity: 99% CAS No.: 687-47-8 Molecular Formula: C5H10O3 Formula Weight: 118.13 Flash Point: 46°(114°F) Physical Form: Liquid Density: 1.036 Refractive Index: 1.4130 Appearance: Colorless Melting Point: -25° Boiling Point: 152-155° Beilstein Reference: 1720839 Merck Reference: 14,3817 Optical Rotation: -11° (neat) UN No.: 1192 MDL No.: MFCD00065359

Formula: C₅H₁₀O₃
MW: 118.13 g/mol
Boiling Pt: 154
Melting Pt: -25
Density: 1.036
Flash Pt: 46 °C (114 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004518
CAS Number: 687-47-8
EINECS: 211-694-1
UN: 1192
ADR: 3 III
Merck Index: 14,3817

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR