2-Ethylhexane-1,3-diol (mixture of isomers) 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Ethoexadiol, Octylene glycol

A10898-0D A10898-0F A10898-AP
CAAAA10898-0DEA 1017.52 CAD
CAAAA10898-0D CAAAA10898-0F CAAAA10898-AP
2-Ethylhexane-1,3-diol (mixture of isomers) 97%
2-Ethylhexane-1,3-diol (mixture of isomers)

MDL: MFCD00004578


Purity: 97% CAS No.: 94-96-2 Molecular Formula: C8H18O2 Formula Weight: 146.23 Odor: Odorless Melting Point: -40° Boiling Point: 241-249° MDL No.: MFCD00004578

Formula: C₈H₁₈O₂
MW: 146.23 g/mol
Boiling Pt: 241…249 °C
Melting Pt: –40 °C
Density: 0.9422 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004578
CAS Number: 94-96-2
EINECS: 202-377-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR