Methyl-m-toluate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10893-18
A10893-30
CAAAA10893-0BEA
1423.23
CAD
CAAAA10893-0B
CAAAA10893-18
CAAAA10893-30
Methyl-m-toluate 98%
Methyl m-toluate
MDL: MFCD00008436
Beilstein Registry No.: 2043574
Purity: 98%
CAS No.: 99-36-5
Molecular Formula: C9H10O2
Formula Weight: 150.18
Flash Point: 95°(203°F)
Refractive Index: 1.5160
Boiling Point: 113°/27mm
Beilstein Reference: 2043574
MDL No.: MFCD00008436
Formula:
C₉H₁₀O₂ Boiling Pt: 113 °C/27 mm Density: 1,062 Flash Pt: 95 °C(203 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008436 CAS Number: 99-36-5 EINECS: 202-753-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...