α,4-Dibromotoluene 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A10892-14
A10892-22
CAAAA10892-14EA
77.42
CAD
CAAAA10892-14
CAAAA10892-22
α,4-Dibromotoluene 98+%
α,4-Dibromotoluene
MDL: MFCD00000179
Beilstein Registry No.: 606498
Purity: 98+%
CAS No.: 589-15-1
Molecular Formula: C7H6Br2
Formula Weight: 249.94
Melting Point: 60-64°
Boiling Point: 115-124°/12mm
Merck Reference: 14,1409
UN No.: 1759
MDL No.: MFCD00000179
Formula:
C₇H₆Br₂ Boiling Pt: 115…124 °C/12 mm Melting Pt: 60…64 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000179 CAS Number: 589-15-1 EINECS: 209-636-5 UN: 1759 ADR: 8,III Merck Index: 14,1409 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...