Diphenylsilane 97%

Supplier: Thermo Scientific Chemicals
Warning

A10884-06 A10884-14 A10884-22
CAAAA10884-06EA 59.09 CAD
CAAAA10884-06 CAAAA10884-14 CAAAA10884-22
Diphenylsilane 97%
Diphenylsilane

MDL: MFCD00003002 Beilstein Registry No.: 2935887 Fieser: 1,348 13,126 15,151,168 16,160 17,132 18,154


Purity: 97% CAS No.: 775-12-2 Molecular Formula: C12H12Si Formula Weight: 184.32 Flash Point: 98°(208°F) Refractive Index: 1.5790 Boiling Point: 95-97°/13mm Beilstein Reference: 2935887 MDL No.: MFCD00003002

Formula: C₁₂H₁₂Si
MW: 184.31 g/mol
Boiling Pt: 95…97 °C/13 mm
Density: 0,996
Flash Pt: 98 °C(208 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003002
CAS Number: 775-12-2
EINECS: 212-271-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR