Methyl toluene-4-sulfonate 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Methyl 4-methylbenzenesulphonate, p-Toluenesulphonic acid methyl ester, Toluene-4-sulfonic acid methyl ester, Methyl p-toluenesulfonate, p-Toluenesulfonic acid methyl ester, Methyl p-toluenesulphonate, Toluene-4-sulphonic acid methyl ester, Methyl 4-methylbenzenesulfonate
MDL: MFCD00008417
Beilstein Registry No.: 609209
Purity: 98%
CAS No.: 80-48-8
Molecular Formula: C8H10O3S
Formula Weight: 186.23
Storage Temperature: 2°C to 8°C
Flash Point: 152°(305°F)
Refractive Index: 1.5185
Melting Point: 24-28°
Boiling Point: 140°/20mm
Beilstein Reference: 609209
UN No.: 3261
MDL No.: MFCD00008417
Formula:
C₈H₁₀O₃S Boiling Pt: 140 °C/20 mm Melting Pt: 24…28 °C Density: 1.234 Flash Pt: 152 °C (305 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00008417 CAS Number: 80-48-8 EINECS: 201-283-5 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...