Methyl-4-hydroxy-3-nitrobenzoate 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10880-18
A10880-30
CAAAA10880-0BEA
2194.79
CAD
CAAAA10880-0B
CAAAA10880-18
CAAAA10880-30
Methyl-4-hydroxy-3-nitrobenzoate 98+%
Methyl 4-hydroxy-3-nitrobenzoate
MDL: MFCD00014695
Beilstein Registry No.: 2115197
Purity: 98+%
CAS No.: 99-42-3
Molecular Formula: C8H7NO5
Formula Weight: 197.15
Melting Point: 73-77°
Beilstein Reference: 2115197
Formula:
C₈H₇NO₅ Melting Pt: 73…77 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014695 CAS Number: 99-42-3 EINECS: 202-755-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...