5-Iodo-α,α,α,α',α',α'-hexafluoro-m-xylene 97+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
5-Iodo-α,α,α,β,β,β-Hexafluoro-m-xylene, 1,3-Di(trifluormethyl)-5-iodbenzene, 1,3-Bis(trifluoromethyl)-5-iodobenzene, 1-Iodo-3,5-bis(trifluoromethyl)benzene, 3,5-Bis(trifluoromethyl)iodobenzene
A10878-06
A10878-14
CAAAA10878-06EA
93.22
CAD
CAAAA10878-06
CAAAA10878-14
CAAAA10878-22
5-Iodo-α,α,α,α',α',α'-hexafluoro-m-xylene 97+%
5-Iodo-α,α,α,α',α',α'-Hexafluoro-m-xylene
MDL: MFCD00040837
Beilstein Registry No.: 2461627
Purity: 97+%
CAS No.: 328-73-4
Molecular Formula: C8H3F6I
Formula Weight: 340.01
Flash Point: 74°(165°F)
Refractive Index: 1.4645
Boiling Point: 62-64°/12mm
Beilstein Reference: 2461627
MDL No.: MFCD00040837
Formula:
C₈H₃F₆I Boiling Pt: 62…64 °C/12 mm Density: 1.919 Flash Pt: 74 °C(165 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00040837 CAS Number: 328-73-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...