Propyl-3,4,5-trihydroxybenzoate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10877-06
A10877-22
A10877-36
CAAAAA10877-06EA
53.48
CAD
CAAAAA10877-06
CAAAAA10877-0E
CAAAA10877-22
CAAAA10877-36
Propyl-3,4,5-trihydroxybenzoate 98%
Propyl 3,4,5-trihydroxybenzoate
MDL: MFCD00002196
Beilstein Registry No.: 1877976
Formula:
C₁₀H₁₂O₅ Boiling Pt: 449 °C (1013 hPa) Melting Pt: 146…150 °C Density: 1.21 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002196 CAS Number: 121-79-9 EINECS: 204-498-2 Merck Index: 14,7859 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...