3,5-Dibromopyridine 98+%

Supplier: Thermo Scientific Chemicals
Warning

A10875-06 A10875-14 A10875-22
CAAAA10875-06EA 109.02 CAD
CAAAA10875-06 CAAAA10875-14 CAAAA10875-22
3,5-Dibromopyridine 98+%
3,5-Dibromopyridine

MDL: MFCD00014634 Beilstein Registry No.: 108477


Purity: 98+% CAS No.: 625-92-3 Molecular Formula: C5H3Br2N Formula Weight: 236.90 Melting Point: 110-113° Beilstein Reference: 108477

Formula: C₅H₃Br₂N
Boiling Pt: 222 °C (1013 hPa)
Melting Pt: 110…113 °C
Storage Temperature: Ambient
MDL Number: MFCD00014634
CAS Number: 625-92-3
EINECS: 210-916-4
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR