Pyridinium chloride 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10871-0B
A10871-18
A10871-30
CAAAA10871-0BEA
469.26
CAD
CAAAA10871-0B
CAAAA10871-18
CAAAA10871-30
Pyridinium chloride 98%
Pyridinium chloride
MDL: MFCD00012802
Beilstein Registry No.: 3615340
Fieser: 1,964 2,352 3,239 4,415
Purity: 98%
CAS No.: 628-13-7
Molecular Formula: C5H5N.HCl
Formula Weight: 115.56
Melting Point: ca 145°
Boiling Point: 222-224°
MDL No.: MFCD00012802
Formula:
C₅H₆ClN Boiling Pt: 222…224 °C Melting Pt: ∼ 145 °C Density: 1.34 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00012802 CAS Number: 628-13-7 EINECS: 211-027-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...