2,5-Dimethylthiophene 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10869-14
A10869-09
CAAAAA10869-14EA
147.26
CAD
CAAAAA10869-14
CAAAA10869-09
2,5-Dimethylthiophene 98+%
2,5-Dimethylthiophene
MDL: MFCD00005452
Beilstein Registry No.: 106450
Purity: 98+%
CAS No.: 638-02-8
Molecular Formula: C6H8S
Formula Weight: 112.19
Flash Point: 24°(75°F)
Refractive Index: 1.5120
Odor: Characteristic
Melting Point: -63°
Boiling Point: 134-136°
Beilstein Reference: 106450
UN No.: 1993
MDL No.: MFCD00005452
Formula:
C₆H₈S Boiling Pt: 134…136 °C Melting Pt: –63 °C Density: 0.984 Flash Pt: 24 °C (75 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005452 CAS Number: 638-02-8 EINECS: 211-313-9 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...