1,8-Diiodooctane 97+% stabilized

Supplier: Thermo Scientific Chemicals
Warning


Stabilization: Stabilised with copper

A10867-14 A10867-18
CAAAAA10867-14EA 225.94 CAD
CAAAAA10867-14 CAAAA10867-18
1,8-Diiodooctane 97+% stabilized
1,8-Diiodooctane

MDL: MFCD00001106 Beilstein Registry No.: 1735437


Purity: 97+% CAS No.: 24772-63-2 Molecular Formula: I(CH2)8I Formula Weight: 366.03 Flash Point: 110°(230°F) Refractive Index: 1.5635 Boiling Point: 167-169°/6mm Beilstein Reference: 1735437 MDL No.: MFCD00001106

Formula: C₈H₁₆I₂
Boiling Pt: 167…169 °C/6 mm
Melting Pt: 16…21 °C
Density: 1.840
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001106
CAS Number: 24772-63-2
EINECS: 246-456-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR