1-Bromo-3-fluorobenzene 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10858-14
A10858-22
A10858-18
A10858-30
CAAAAA10858-14EA
79.95
CAD
CAAAAA10858-14
CAAAAA10858-22
CAAAA10858-18
CAAAA10858-30
1-Bromo-3-fluorobenzene 99%
1-Bromo-3-fluorobenzene
MDL: MFCD00000326
Beilstein Registry No.: 1853969
Purity: 99%
CAS No.: 1073-06-9
Molecular Formula: C6H4BrF
Formula Weight: 175.01
Flash Point: 46°(115°F)
Refractive Index: 1.5275
Odor: Aromatic
Melting Point: -8°
Boiling Point: 148-150°
Beilstein Reference: 1853969
UN No.: 1993
MDL No.: MFCD00000326
Formula:
C₆H₄BrF Boiling Pt: 148…150 °C Melting Pt: –8 °C Density: 1.594 Flash Pt: 46 °C (115 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000326 CAS Number: 1073-06-9 EINECS: 214-023-0 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...