3-Chlor-4-fluorothiophenol 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10856-03
A10856-06
CAAAA10856-03EA
138.72
CAD
CAAAA10856-03
CAAAA10856-06
3-Chlor-4-fluorothiophenol 97%
3-Chlor-4-fluorothiophenol
MDL: MFCD00052938
Purity: 97%
CAS No.: 60811-23-6
Molecular Formula: C6H4ClFS
Formula Weight: 162.61
Boiling Point: 68-70°/2mm
MDL No.: MFCD00052938
Formula:
C₆H₄ClFS Boiling Pt: 68…70 °C/2 mm Storage Temperature: Ambient |
MDL Number:
MFCD00052938 CAS Number: 60811-23-6 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...