4-Bromo-N,N-dimethylaniline 98+%

Supplier: Thermo Scientific Chemicals
Warning

A10854-14 A10854-22
CAAAA10854-14EA 60.04 CAD
CAAAA10854-14 CAAAA10854-22
4-Bromo-N,N-dimethylaniline 98+%
4-Bromo-N,N-dimethylaniline

MDL: MFCD00000093 Beilstein Registry No.: 2206155


Purity: 98+% CAS No.: 586-77-6 Molecular Formula: C8H10BrN Formula Weight: 200.08 Flash Point: 110°(230°F) Melting Point: 52-56° Boiling Point: 263-265° Beilstein Reference: 2206155 MDL No.: MFCD00000093

Formula: C₈H₁₀BrN
Boiling Pt: 263…265 °C
Melting Pt: 52…56 °C
Density: 1.3218 g/cm³ (99.5 °C)
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000093
CAS Number: 586-77-6
EINECS: 209-582-2
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR