Benzenesulfonyl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Benzene sulfochloride, Benzene sulphochloride

A10849-0B A10849-30
CAAAA10849-0BEA 150.57 CAD
CAAAA10849-0B CAAAA10849-0I CAAAA10849-30
Benzenesulfonyl chloride 98%
Benzenesulphonyl chloride

MDL: MFCD00007426 Beilstein Registry No.: 606926 Fieser: 1,46 5,22


Purity: 98% CAS No.: 98-09-9 Molecular Formula: C6H5ClO2S Formula Weight: 176.62 Flash Point: 132°(270°F) Refractive Index: 1.5525 Odor: Irritating Melting Point: 14-15° Boiling Point: 252° Beilstein Reference: 606926 Merck Reference: 14,1072 UN No.: 2225 MDL No.: MFCD00007426

Formula: C₆H₅ClO₂S
Boiling Pt: 252 °C
Melting Pt: 14…15 °C
Density: 1.377
Flash Pt: 132 °C (270 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007426
CAS Number: 98-09-9
EINECS: 202-636-6
UN: 2225
ADR: 8,III
Merck Index: 14,1072

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR