2-Picoline 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Methylpyridine, α-Picoline
A10847-0F
A10847-AE
A10847-AP
CAAAA10847-0FEA
213.3
CAD
CAAAA10847-0F
CAAAA10847-AE
CAAAA10847-AP
2-Picoline 98+%
2-Picoline
MDL: MFCD00006332
Beilstein Registry No.: 104581
Purity: 98+%
CAS No.: 109-06-8
Molecular Formula: C6H7N
Formula Weight: 93.13
Flash Point: 26°(78°F)
Refractive Index: 1.5000
Melting Point: -70°
Boiling Point: 128-129°
Beilstein Reference: 104581
Merck Reference: 14,7400
UN No.: 2313
MDL No.: MFCD00006332
Formula:
C₆H₇N MW: 93.13 g/mol Boiling Pt: 128…129 °C Melting Pt: –70 °C Density: 0.944 Flash Pt: 26 °C (78 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006332 CAS Number: 109-06-8 EINECS: 203-643-7 UN: 1992 ADR: 3,III Merck Index: 14,7400 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...