Dibenzyl malonate 95%

Supplier: Thermo Scientific Chemicals

Synonyms: Malonic acid dibenzyl ester, 1,3-Propanedioic acid dibenzyl ester

A10844-09 A10844-18 A10844-30
CAAAA10844-09EA 104.91 CAD
CAAAA10844-09 CAAAA10844-18 CAAAA10844-30
Dibenzyl malonate 95%
Dibenzyl malonate

MDL: MFCD00004779 Beilstein Registry No.: 1998264 Fieser: 1,198


Purity: 95% CAS No.: 15014-25-2 Molecular Formula: C17H16O4 Formula Weight: 284.31 Flash Point: 110°(230°F) Refractive Index: 1.5440 Boiling Point: 188°/0.2mm Beilstein Reference: 1998264 MDL No.: MFCD00004779

Formula: C₁₇H₁₆O₄
MW: 284.31 g/mol
Boiling Pt: 188
Density: 1.137
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004779
CAS Number: 15014-25-2
EINECS: 239-099-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR