3-Acetylbenzonitrile 97+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10841-06
A10841-14
CAAAA10841-06EA
111.23
CAD
CAAAA10841-06
CAAAA10841-14
CAAAA10841-22
3-Acetylbenzonitrile 97+%
3-Acetylbenzonitrile
MDL: MFCD00001806
Beilstein Registry No.: 2079629
Purity: 97+%
CAS No.: 6136-68-1
Molecular Formula: C9H7NO
Formula Weight: 145.16
Melting Point: 97-101°
Boiling Point: 120°/5mm
UN No.: 3439
MDL No.: MFCD00001806
Formula:
C₉H₇NO Boiling Pt: 120 °C/5 mm Melting Pt: 97…101 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001806 CAS Number: 6136-68-1 EINECS: 228-110-6 UN: 3439 ADR: "6.1",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...