3-Fluoro-4-nitrotoluene 99%

Supplier: Thermo Scientific Chemicals
Warning

A10840-06 A10840-14
CAAAA10840-06EA 49.45 CAD
CAAAA10840-06 CAAAA10840-14
3-Fluoro-4-nitrotoluene 99%
3-Fluoro-4-nitrotoluene

MDL: MFCD00007053 Beilstein Registry No.: 2502584


Purity: 99% CAS No.: 446-34-4 Molecular Formula: C7H6FNO2 Formula Weight: 155.13 Flash Point: 110°(230°F) Melting Point: 51-55° Boiling Point: 97-98°/3mm Beilstein Reference: 2502584 UN No.: 2811 MDL No.: MFCD00007053

Formula: C₇H₆FNO₂
Boiling Pt: 97…98 °C/3 mm
Melting Pt: 51…55 °C
Density: 1.438
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007053
CAS Number: 446-34-4
EINECS: 207-166-5
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR