3-Ethyl-2-iodotoluene 98%
Supplier: Thermo Scientific Chemicals
A10839-06
A10839-14
CAAAA10839-06EA
98.91
CAD
CAAAA10839-06
CAAAA10839-14
3-Ethyl-2-iodotoluene 98%
3-Ethyl-2-iodotoluene
MDL: MFCD00052855
Purity: 98%
CAS No.: 175277-95-9
Molecular Formula: C9H11I
Formula Weight: 246.08
Boiling Point: 118-120°/15mm
MDL No.: MFCD00052855
Formula:
C₉H₁₁I Boiling Pt: 118…120 °C/15 mm Storage Temperature: Ambient |
MDL Number:
MFCD00052855 CAS Number: 175277-95-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...