4'-Hydroxychalcone 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-(4-Hydroxyphenyl)-3-phenylprop-2-en-1-one, 4'-Hydroxybenzylidenacetophenone

A10838-06 A10838-14
CAAAA10838-06EA 84.06 CAD
CAAAA10838-06 CAAAA10838-14
4'-Hydroxychalcone 97%
4'-Hydroxychalcone

MDL: MFCD00016484 Beilstein Registry No.: 2049150


Purity: 97% CAS No.: 2657-25-2 Molecular Formula: C15H12O2 Formula Weight: 224.26 Melting Point: 175-177° Beilstein Reference: 2049150

Formula: C₁₅H₁₂O₂
MW: 224.26 g/mol
Melting Pt: 175…177 °C
Storage Temperature: Ambient
MDL Number: MFCD00016484
CAS Number: 2657-25-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR