ω,2,4-Trichloroacetophenone 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2,4-Dichlorophenacyl chloride, α,2,4-Trichloroacetophenone, 2,2',4'-Trichloroacetophenone
A10835-22
A10835-36
CAAAA10835-22EA
155
CAD
CAAAA10835-22
CAAAA10835-36
ω,2,4-Trichloroacetophenone 97%
ω,2,4-Trichloroacetophenone
MDL: MFCD00000934
Beilstein Registry No.: 957098
Purity: 97%
CAS No.: 4252-78-2
Molecular Formula: C8H5Cl3O
Formula Weight: 223.49
Flash Point:
Formula:
C₈H₅Cl₃O Boiling Pt: 292 °C (1013 hPa) Melting Pt: 52…56 °C Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000934 CAS Number: 4252-78-2 EINECS: 224-218-2 UN: 2923 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...