2-(2-Aminoethylamino)ethanol 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: N-(2-Hydroxyethyl)ethylenediamine, AEEA

A10833-22 A10833-0E A10833-36
CAAAAA10833-22EA 30.18 CAD
CAAAAA10833-22 CAAAA10833-0E CAAAA10833-36
2-(2-Aminoethylamino)ethanol 99%
2-(2-Aminoethylamino)ethanol

MDL: MFCD00008170 Beilstein Registry No.: 506012


Purity: 99% CAS No.: 111-41-1 Molecular Formula: C4H12N2O Formula Weight: 104.15 Flash Point: 125°(257°F) Refractive Index: 1.4861 Odor: Amine-like pH: 11.8 Melting Point: -28° Boiling Point: 238-240° Beilstein Reference: 506012 UN No.: 2735 MDL No.: MFCD00008170

Formula: C₄H₁₂N₂O
MW: 104.15 g/mol
Boiling Pt: 238…240 °C
Melting Pt: –28 °C
Density: 1.029
Flash Pt: 125 °C (257 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008170
CAS Number: 111-41-1
EINECS: 203-867-5
UN: 2735
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR