2-Chlorobenzoic acid 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10832-0B
A10832-30
CAAAA10832-0BEA
134.77
CAD
CAAAA10832-0B
CAAAA10832-0I
CAAAA10832-30
2-Chlorobenzoic acid 98+%
2-Chlorobenzoic acid
MDL: MFCD00002412
Beilstein Registry No.: 907340
Purity: 98+%
CAS No.: 118-91-2
Molecular Formula: C7H5ClO2
Formula Weight: 156.57
Flash Point: 173°(343°F)
Melting Point: 138-142°
Boiling Point: 284-286°
Beilstein Reference: 907340
Merck Reference: 14,2124
MDL No.: MFCD00002412
Formula:
C₇H₅ClO₂ Boiling Pt: 284…286 °C Melting Pt: 138…142 °C Density: 1.544 Flash Pt: 173 °C(343 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002412 CAS Number: 118-91-2 EINECS: 204-285-4 Merck Index: 14,2124 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...