Hydroxyurea 98%, REacton®

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: N-(Aminocarbonyl)hydroxylamine, Hydroxycarbamide

A10831-14 A10831-03 A10831-06
CAAAAA10831-14EA 639.9 CAD
CAAAAA10831-14 CAAAA10831-03 CAAAA10831-06
Hydroxyurea 98%, REacton®
Hydroxyurea

An inhibitor of DNA synthesis and an anticancer agent


Purity: 98% CAS No.: 127-07-1 Molecular Formula: CH4N2O2 Formula Weight: 76.06 Melting Point: ca 140° dec. Beilstein Reference: 1741548 Merck Reference: 14,4848 MDL No.: MFCD00007943


An inhibitor of DNA synthesis and an anticancer agent

Formula: CH₄N₂O₂
MW: 76.05 g/mol
Boiling Pt: 296 °C (1013 hPa)
Melting Pt: ∼ 140 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00007943
CAS Number: 127-07-1
EINECS: 204-821-7
Merck Index: 14,4848

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR