(±)-1-Phenyl-1-propanol 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-Phenyl-1-propanol, (±)-α-Ethylbenzyl alcohol
A10830-14
A10830-22
CAAAA10830-14EA
42.82
CAD
CAAAA10830-14
CAAAA10830-22
CAAAA10830-36
(±)-1-Phenyl-1-propanol 98+%
(±)-1-Phenyl-1-propanol
MDL: MFCD00004564
Beilstein Registry No.: 1906759
Purity: 98+%
CAS No.: 93-54-9
Molecular Formula: C9H12O
Formula Weight: 136.19
Flash Point: 95°(203°F)
Refractive Index: 1.5200
Boiling Point: 217-219°
Beilstein Reference: 1906759
Merck Reference: 14,3768
MDL No.: MFCD00004564
Formula:
C₆H₅CH(OH)CH₂CH₃ MW: 136.19 g/mol Boiling Pt: 217…219 °C Density: 0.994 Flash Pt: 95 °C(203 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004564 CAS Number: 93-54-9 EINECS: 202-256-0 Merck Index: 14,3768 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...