1,3-Propanediol 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
1,3-Propylene glycol, 1,3-Dihydroxy propane
A10829-0B
A10829-0I
A10829-30
CAAAA10829-0BEA
402.9
CAD
CAAAA10829-0B
CAAAA10829-0I
CAAAA10829-30
1,3-Propanediol 99%
1,3-Propanediol
MDL: MFCD00002949
Beilstein Registry No.: 969155
Miscible with water, alcohol
Purity: 99%
CAS No.: 504-63-2
Molecular Formula: HO(CH2)3OH
Formula Weight: 76.10
Flash Point: 140°(174°F)
Refractive Index: 1.4400
Melting Point: -26°
Boiling Point: 214°
Solubility: Miscible with water, alcohol
Beilstein Reference: 969155
Merck Reference: 14,9714
MDL No.: MFCD00002949
Formula:
HO(CH₂)₃OH MW: 76.1 g/mol Boiling Pt: 214 Melting Pt: -32 Density: 1.053 Flash Pt: 140 °C (174 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002949 CAS Number: 504-63-2 EINECS: 207-997-3 Merck Index: 14,9714 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...