2-Amino-4-methylbenzothiazole 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Methylbenzothiazol-2-ylamine, 4-Methylbenzo[d]thiazol-2-amine

A10822-22 A10822-06 A10822-14
CAAAAA10822-22EA 259.12 CAD
CAAAAA10822-22 CAAAA10822-06 CAAAA10822-14
2-Amino-4-methylbenzothiazole 98%
2-Amino-4-methylbenzothiazole

MDL: MFCD00005793 Beilstein Registry No.: 129346


Purity: 98% CAS No.: 1477-42-5 Molecular Formula: C8H8N2S Formula Weight: 164.22 Melting Point: 137-139° Beilstein Reference: 129346

Formula: C₈H₈N₂S
Boiling Pt: 321 °C (1013 hPa)
Melting Pt: 137…139 °C
Storage Temperature: Ambient
MDL Number: MFCD00005793
CAS Number: 1477-42-5
EINECS: 216-028-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR