m-Anisidine 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
3-Aminoanisole, 3-Methoxyaniline, 3-Anisidine
A10815-0B
A10815-18
A10815-30
CAAAA10815-0BEA
605.14
CAD
CAAAA10815-0B
CAAAA10815-18
CAAAA10815-30
m-Anisidine 98%
m-Anisidine
MDL: MFCD00007783
Beilstein Registry No.: 386119
Purity: 98%
CAS No.: 536-90-3
Molecular Formula: C7H9NO
Formula Weight: 123.16
Flash Point: 126°(258°F)
Refractive Index: 1.5810
Odor: Characteristic
Melting Point: -1°
Boiling Point: 250-251°
Beilstein Reference: 386119
Merck Reference: 14,667
UN No.: 2431
MDL No.: MFCD00007783
Formula:
C₇H₉NO Boiling Pt: 250…251 °C Melting Pt: –1 °C Density: 1.101 Flash Pt: 126 °C (258 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007783 CAS Number: 536-90-3 EINECS: 208-651-4 UN: 2431 ADR: 6.1,III Merck Index: 14,667 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...