Levulinic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10813-0B
A10813-30
CAAAA10813-0BEA
206.98
CAD
CAAAA10813-0B
CAAAA10813-30
Levulinic acid 98%
Levulinic acid
MDL: MFCD00002796
Beilstein Registry No.: 506796
Fieser: 1,564 2,239 10,230
Purity: 98%
CAS No.: 123-76-2
Molecular Formula: C5H8O3
Formula Weight: 116.12
Flash Point: 137°(278°F)
Refractive Index: 1.4415
Odor: Characteristic
pH: 2.5-3.0
Melting Point: 26-31°
Boiling Point: 245-246°
Beilstein Reference: 506796
Merck Reference: 14,5472
UN No.: 3261
MDL No.: MFCD00002796
Formula:
C₅H₈O₃ Boiling Pt: 245…246 °C Melting Pt: 26…31 °C Density: 1.134 Flash Pt: 137 °C(278 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002796 CAS Number: 123-76-2 EINECS: 204-649-2 UN: 3261 ADR: 8,III Merck Index: 14,5472 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...