4-Bromo-2-fluorophenol 97+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Bromo-3-fluoro-4-hydroxybenzene

A10804-06 A10804-14 A10804-22
CAAAA10804-06EA 34.92 CAD
CAAAA10804-06 CAAAA10804-14 CAAAA10804-22
4-Bromo-2-fluorophenol 97+%
4-Bromo-2-fluorophenol

MDL: MFCD00011722 Beilstein Registry No.: 1861281


Purity: 97+% CAS No.: 2105-94-4 Molecular Formula: C6H4BrFO Formula Weight: 191.00 Flash Point: 98°(208°F) Refractive Index: 1.5660 Boiling Point: 82-83°/10mm Beilstein Reference: 1861281 MDL No.: MFCD00011722

Formula: C₆H₄BrFO
MW: 191 g/mol
Boiling Pt: 82…83 °C/10 mm
Density: 1.744
Flash Pt: 98 °C (208 °F)
Storage Temperature: Ambient
MDL Number: MFCD00011722
CAS Number: 2105-94-4
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR