Acetone oxime 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Propan-2-one oxime, Acetoxime, Acetone oxime, 2-Propanone oxime
A10802-0B
A10802-18
A10802-30
CAAAA10802-0BEA
450.3
CAD
CAAAA10802-0B
CAAAA10802-18
CAAAA10802-30
Acetone oxime 98%
Acetone oxime
MDL: MFCD00002118
Beilstein Registry No.: 1560146
Purity: 98%
CAS No.: 127-06-0
Molecular Formula: C3H7NO
Formula Weight: 73.10
Flash Point: 60°(140°F)
Melting Point: 60-63°
Boiling Point: 133-135°
Beilstein Reference: 1560146
Merck Reference: 14,75
UN No.: 1325
MDL No.: MFCD00002118
Formula:
C₃H₇NO MW: 73.09 g/mol Boiling Pt: 133…135 °C Melting Pt: 60…63 °C Density: 0.901 Flash Pt: 60 °C (140 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002118 CAS Number: 127-06-0 EINECS: 204-820-1 UN: 1325 ADR: 4.1 III Merck Index: 14,75 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...