4-Fluoro-3-nitrobenzoic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10800-22
A10800-06
A10800-14
CAAAAA10800-22EA
733.12
CAD
CAAAAA10800-22
CAAAA10800-06
CAAAA10800-14
4-Fluoro-3-nitrobenzoic acid 98%
4-Fluoro-3-nitrobenzoic acid
MDL: MFCD00007058
Beilstein Registry No.: 2107562
Purity: 98%
CAS No.: 453-71-4
Molecular Formula: C7H4FNO4
Formula Weight: 185.11
Melting Point: 121-126°
Boiling Point: 92°/15mm
MDL No.: MFCD00007058
Formula:
C₇H₄FNO₄ Boiling Pt: 92 °C/15 mm Melting Pt: 121…126 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007058 CAS Number: 453-71-4 EINECS: 207-221-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...