1,3-Difluoro-4-iodobenzene 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,4-Difluoroiodobenzene, 2,4-Difluoro-1-iodobenzene

A10798-09 A10798-18
CAAAA10798-09EA 67.62 CAD
CAAAA10798-09 CAAAA10798-18 CAAAA10798-30
1,3-Difluoro-4-iodobenzene 99%
1,3-Difluoro-4-iodobenzene

MDL: MFCD00040151 Beilstein Registry No.: 2081078


Purity: 99% CAS No.: 2265-93-2 Molecular Formula: C6H3F2I Formula Weight: 239.99 Flash Point: 67°(152°F) Refractive Index: 1.5580 Boiling Point: 175-176° Beilstein Reference: 2081078 MDL No.: MFCD00040151

Formula: C₆H₃F₂I
MW: 239.99 g/mol
Boiling Pt: 175…176 °C
Density: 2.006
Flash Pt: 67 °C (152 °F)
Storage Temperature: Ambient
MDL Number: MFCD00040151
CAS Number: 2265-93-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR