Salicylamide 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10797-0B
A10797-30
CAAAA10797-0BEA
143.15
CAD
CAAAA10797-0B
CAAAA10797-0I
CAAAA10797-30
Salicylamide 98%
Salicylamide
MDL: MFCD00007978
Beilstein Registry No.: 742439
Purity: 98%
CAS No.: 65-45-2
Molecular Formula: C7H7NO2
Formula Weight: 137.14
Melting Point: 138-142°
Boiling Point: 181°/14mm
Merck Reference: 14,8328
MDL No.: MFCD00007978
Formula:
C₇H₇NO₂ Boiling Pt: 181 °C/14 mm Melting Pt: 138…142 °C Density: 1.33 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00007978 CAS Number: 65-45-2 EINECS: 200-609-3 Merck Index: 14,8328 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...