4,5-Dimethylthiazole 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10785-06
A10785-14
A10785-22
CAAAA10785-06EA
44.56
CAD
CAAAA10785-06
CAAAA10785-14
CAAAA10785-22
4,5-Dimethylthiazole 98%
4,5-Dimethylthiazole
MDL: MFCD00005336
Beilstein Registry No.: 105694
Purity: 98%
CAS No.: 3581-91-7
Molecular Formula: C5H7NS
Formula Weight: 113.18
Flash Point: 51°(123°F)
Refractive Index: 1.5210
Boiling Point: 157-158°
Beilstein Reference: 105694
UN No.: 1993
MDL No.: MFCD00005336
Formula:
C₅H₇NS Boiling Pt: 157…158 °C Density: 1.070 Flash Pt: 51 °C (123 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005336 CAS Number: 3581-91-7 EINECS: 222-703-3 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...