α-Chloro-m-xylene 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
1-(Chloromethyl)-3-methylbenzene, 3-Methylbenzyl chloride
A10784-22
A10784-36
CAAAA10784-22EA
165.9
CAD
CAAAA10784-22
CAAAA10784-36
α-Chloro-m-xylene 98%
α-Chloro-m-xylene
MDL: MFCD00000909
Beilstein Registry No.: 2040357
Purity: 98%
CAS No.: 620-19-9
Molecular Formula: C8H9Cl
Formula Weight: 140.61
Flash Point: 75°(167°F)
Refractive Index: 1.5350
Odor: Pungent
Boiling Point: 195-196°
Beilstein Reference: 2040357
Merck Reference: 14,10091
UN No.: 3265
MDL No.: MFCD00000909
Formula:
C₈H₉Cl MW: 140.61 g/mol Boiling Pt: 195…196 °C Density: 1.066 Flash Pt: 75 °C (167 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000909 CAS Number: 620-19-9 EINECS: 210-628-9 UN: 3265 ADR: 8,III Merck Index: 14,10091 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...