4-Octylbenzoic acid 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10777-03
A10777-06
CAAAA10777-03EA
102.38
CAD
CAAAA10777-03
CAAAA10777-06
4-Octylbenzoic acid 99%
4-Octylbenzoic acid
MDL: MFCD00042649
Beilstein Registry No.: 1954607
Purity: 99%
CAS No.: 3575-31-3
Molecular Formula: C15H22O2
Formula Weight: 234.34
Melting Point: 97-98°
Beilstein Reference: 1954607
Formula:
C₁₅H₂₂O₂ Boiling Pt: 362 °C (1013 hPa) Melting Pt: 97…98 °C Storage Temperature: Ambient |
MDL Number:
MFCD00042649 CAS Number: 3575-31-3 EINECS: 222-692-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...