Triethylene glycol 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
2,2'-(Ethylenedioxy)diethanol, 2,2'-(Ethane-1,2-diylbis(oxy))diethanol
MDL: MFCD00002880
Beilstein Registry No.: 969357
Purity: 99%
CAS No.: 112-27-6
Molecular Formula: C6H14O4
Formula Weight: 150.17
Flash Point: 165°(329°F)
Physical Form: Liquid
Density: 1.125
Refractive Index: 1.4555
Sensitivity: Hygroscopic
Appearance: Colorless
Odor: Nearly odorless
Melting Point: -7°
Boiling Point: 284-288°
Beilstein Reference: 969357
Merck Reference: 14,9670
MDL No.: MFCD00002880
Formula:
C₆H₁₄O₄ MW: 150.17 g/mol Boiling Pt: 285 Melting Pt: -7 Density: 1.125 Flash Pt: 165 °C (329 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002880 CAS Number: 112-27-6 EINECS: 203-953-2 Merck Index: 14,9670 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...