o-Anisaldehyde 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Salicylaldehyde methyl ether, 2-Methoxybenzaldehyde, 2-Anisaldehyde

A10770-22 A10770-36
CAAAA10770-0EEA 1391.95 CAD
CAAAA10770-0E CAAAA10770-22 CAAAA10770-36
o-Anisaldehyde 98%
o-Anisaldehyde

MDL: MFCD00003308 Beilstein Registry No.: 606301


Purity: 98% CAS No.: 135-02-4 Molecular Formula: C8H8O2 Formula Weight: 136.15 Flash Point: 117°(242°F) Refractive Index: 1.5610 Odor: Characteristic Melting Point: 35-39° Boiling Point: 237-238° Beilstein Reference: 606301 MDL No.: MFCD00003308

Formula: C₈H₈O₂
MW: 136.15 g/mol
Boiling Pt: 237…238 °C
Melting Pt: 35…39 °C
Density: 1,13
Flash Pt: 117 °C (242 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003308
CAS Number: 135-02-4
EINECS: 205-171-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR