m-Anisic acid 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-Anisic acid, 3-Methoxybenzoic acid
A10767-18
A10767-30
CAAAAA10767-0BEA
1731
CAD
CAAAAA10767-0B
CAAAA10767-18
CAAAA10767-30
m-Anisic acid 99%
m-Anisic acid
MDL: MFCD00002499
Beilstein Registry No.: 508838
Purity: 99%
CAS No.: 586-38-9
Molecular Formula: C8H8O3
Formula Weight: 152.15
Odor: Odorless
Melting Point: 104-108°
Boiling Point: 170-172°/10mm
Beilstein Reference: 508838
MDL No.: MFCD00002499
Formula:
C₈H₈O₃ Boiling Pt: 170…172 °C (10 mmHg) Melting Pt: 104…108 °C Density: 1.14 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002499 CAS Number: 586-38-9 EINECS: 209-574-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...