2-Methylindole 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10764-0B
A10764-18
A10764-30
CAAAA10764-0BEA
894.28
CAD
CAAAA10764-0B
CAAAA10764-18
CAAAA10764-30
2-Methylindole 98+%
2-Methylindole
MDL: MFCD00005616
Beilstein Registry No.: 109781
Purity: 98+%
CAS No.: 95-20-5
Molecular Formula: C9H9N
Formula Weight: 131.18
Flash Point: 141°(285°F)
Melting Point: 56-60°
Boiling Point: 272-273°
Beilstein Reference: 109781
MDL No.: MFCD00005616
Formula:
C₉H₉N Boiling Pt: 272…273 °C Melting Pt: 56…60 °C Density: 1.07 Flash Pt: 141 °C(285 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005616 CAS Number: 95-20-5 EINECS: 202-398-3 UN: 3335 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...