Dodecyltrimethylammonium bromide 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Dodecyl trimethyl ammonium bromide, (1-Dodecyl)trimethylammonium bromide
A10761-09
A10761-18
A10761-30
CAAAA10761-09EA
77.26
CAD
CAAAA10761-09
CAAAA10761-18
CAAAA10761-30
Dodecyltrimethylammonium bromide 99%
Dodecyltrimethylammonium bromide
MDL: MFCD00011767
Beilstein Registry No.: 3597463
Purity: 99%
CAS No.: 1119-94-4
Molecular Formula: CH3(CH2)11N(CH3)3Br
Formula Weight: 308.35
Melting Point: ca 245° dec.
Beilstein Reference: 3597463
UN No.: 3077
MDL No.: MFCD00011767
Formula:
C₁₅H₃₄BrN MW: 308.35 g/mol Melting Pt: ∼ 245 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00011767 CAS Number: 1119-94-4 EINECS: 214-290-3 UN: 3077 ADR: 9 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...