Kojic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Hydroxy-2-hydroxymethyl-4H-pyran-4-one, 5-Hydroxy-2-(hydroxymethyl)-4H-pyran-4-one, 5-Hydroxy-2-hydroxymethyl-4-pyrone, 5-Hydroxy-2-hydroxymethyl-γ-pyrone

A10760-06 A10760-14 A10760-22
CAAAA10760-06EA 87.06 CAD
CAAAA10760-06 CAAAA10760-14 CAAAA10760-22
Kojic acid 99%
Kojic acid

A chelation agent that inhibits melanin production. Also a tyrosine kinase inhibitor


Purity: 99% CAS No.: 501-30-4 Molecular Formula: C6H6O4 Formula Weight: 142.11 Odor: Odorless Melting Point: 152-155° Beilstein Reference: 120895 Merck Reference: 14,5317 MDL No.: MFCD00006580

Formula: C₆H₆O₄
MW: 142.11 g/mol
Boiling Pt: 323 °C (1013 hPa)
Melting Pt: 152…155 °C
Density: 1.58 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006580
CAS Number: 501-30-4
EINECS: 207-922-4
Merck Index: 14,5317

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR