Methyl hexanoate 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Caproic acid methyl ester, Methyl caproate, Hexanoic acid methyl ester
A10757-0B
A10757-18
CAAAA10757-0BEA
458.2
CAD
CAAAA10757-0B
CAAAA10757-18
Methyl hexanoate 99%
Methyl hexanoate
MDL: MFCD00009510
Beilstein Registry No.: 1744683
Purity: 99%
CAS No.: 106-70-7
Molecular Formula: CH3(CH2)4CO2CH3
Formula Weight: 130.19
Flash Point: 45°(113°F)
Refractive Index: 1.4050
Melting Point: -71°
Boiling Point: 150-151°
Beilstein Reference: 1744683
UN No.: 3272
MDL No.: MFCD00009510
Formula:
C₇H₁₄O₂ MW: 130.19 g/mol Boiling Pt: 150…151 °C Melting Pt: –71 °C Density: 0.885 Flash Pt: 45 °C (113 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009510 CAS Number: 106-70-7 EINECS: 203-425-1 UN: 3272 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...