2-Chloro-α,α,α-trifluorotoluene 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-Chloro-2-(trifluoromethyl)benzene, 2-Chlorobenzotrifluoride
A10753-0E
A10753-22
A10753-36
CAAAA10753-0EEA
352.34
CAD
CAAAA10753-0E
CAAAA10753-22
CAAAA10753-36
2-Chloro-α,α,α-trifluorotoluene 99%
2-Chloro-α,α,α-trifluorotoluene
MDL: MFCD00000561
Beilstein Registry No.: 510993
Purity: 99%
CAS No.: 88-16-4
Molecular Formula: C7H4ClF3
Formula Weight: 180.56
Flash Point: 58°(136°F)
Refractive Index: 1.4560
Melting Point: -6°
Boiling Point: 151-153°
Beilstein Reference: 510993
UN No.: 2234
MDL No.: MFCD00000561
Formula:
C₇H₄ClF₃ Boiling Pt: 151…153 °C Melting Pt: –6 °C Density: 1.369 Flash Pt: 58 °C (136 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000561 CAS Number: 88-16-4 EINECS: 201-805-1 UN: 2234 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...